Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Uniflo™ 25 Syringe Filters
Ideal for filtration of organic-based samples. Whatman™ Uniflo Syringe Filter, PTFE Membrane is a disposable, hydrophobic filter unit that will not allow water to pass without high pressures. The polytetrafluoroethylene membrane will also stop aqueous aerosols in gas streams.
Specifications
Housing Material Polypropylene
Membrane Hydrophobic
Sterility Non-sterile
Disposable  Yes
Inlet Style Female Luer Lock
View More Specs
Products 7
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size No. per Pack Price Quantity & Availability  
09-927-580
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463882
13mm 0.2μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-323
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463881
13mm 0.2μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-439
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463413
25mm 0.2μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-330
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463423
25mm 0.45μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-328
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463823
13mm 0.45μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-325
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463880
13mm 0.45μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-429
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463422
25mm 0.45μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Whatman Uniflo Syringe Filters are disposable filter units designed to provide clean filtrate from small volumes up to 10mL (13mm filters) or 100mL (25mm filters).
  • Combines premium quality with economic efficiency
  • Available in a variety of membrane choices with a polypropylene overmold housing
  • A range of sizes and pore sizes to meet the filtration application needs
  • Individual ink-jet printing on the filter for easy identification
  • Packaged in a clean, compact recyclable plastic box to optimize benchtop space
Specifications
Specifications
Polypropylene
Non-sterile
Female Luer Lock
HPLC sample filtration
Resealable, recyclable plastic boxes
Hydrophobic
 Yes
Male Slip Luer
Organic-based samples
UNIFLO Syringe Filter