Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).
Manufacturer:  VITLAB™ 302597
 View more versions of this product
Catalog No. 03-051-002
Add to cart
Description
Description
  • Permanent magnet AlNiCo V core
  • Angled shape gives rise to significant turbulence
  • Achieves effective mixing, even at low spin rates
  • FTE
Specifications
Specifications
For preparation, separation and concentration
PTFE
80mm
5/Pk.
Magnetic stirring bar
Permanent magnet AlNiCo V core
10mm
PTFE (Polytetrafluoroethylene)
Polygonal