Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549717

Catalog No. PA549717


Add to cart

Description

Description

The antibody detects endogenous level of total PTGER1 / EP1 protein.

Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(q) proteins which activate a phosphatidylinositol-calcium second messenger system. May play a role as an important modulator of renal function. Implicated the smooth muscle contractile response to PGE2 in various tissues.
Specifications

Specifications

PTGER1
Polyclonal
Unconjugated
PTGER1
Liquid
PTGER1
Synthetic peptide corresponding to a region derived from 399-402 amino acids of Human prostaglandin E receptor 1 (subtype EP1), 42kDa
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P34995
prostaglandin E receptor 1 (subtype EP1), 42kDa, EP1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5731
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit