Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER1, Mouse anti-Human, Clone: 1014918, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $298.50

Specifications

Antigen PTGER1
Clone 1014918
Immunogen Synthetic peptide containing human PTGER1, Accession # P34995
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1008110
View Documents
R&D Systems
MAB100081100
100 ug
Each for $298.50
Add to cart
 
MAB100081SP
View Documents
R&D Systems
MAB100081SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

PTGER1 Monoclonal antibody specifically detects PTGER1 in Human Samples. It is validated for Immunohistochemistry.
Specifications

Specifications

PTGER1
Synthetic peptide containing human PTGER1, Accession # P34995
Primary
Human
Unconjugated
5731
Purified
Immunohistochemistry
RUO
1014918
Mouse
Monoclonal
IgG2b
EP1, PGE receptor EP1 subtype, PGE receptor, EP1 subtype, PGE2 receptor EP1 subtype, prostaglandin E receptor 1 (subtype EP1), 42kD, prostaglandin E receptor 1 (subtype EP1), 42kDa, prostaglandin E receptor 1, subtype EP1, prostaglandin E2 receptor EP1 subtype, Prostanoid EP1 receptor
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit