Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTGER3 Mouse anti-Bovine, Clone: 5F5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266637
Description
PTGER3 Monoclonal antibody specifically detects PTGER3 in Bovine samples. It is validated for Western Blot.Specifications
PTGER3 | |
5F5 | |
Western Blot | |
PBS with 0.08% Sodium Azide | |
5733 | |
Mouse | |
IgG2a | |
Primary | |
100μg | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Purified | |
EP3EP3e, EP3-I, EP3-II, EP3-III, EP3-IV, MGC141828, MGC141829, MGC27302, PGE receptor EP3 subtype, PGE receptor, EP3 subtype, PGE2 receptor EP3 subtype, PGE2-R, prostaglandin E receotor EP3 subtype 3 isoform, prostaglandin E receptor 3 (subtype EP3), prostaglandin E2 receptor EP3 subtype, prostaglandin receptor (PGE-2), Prostanoid EP3 receptor | |
PTGER3 | |
Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human EP3 receptor protein and mouse myeloma cells. | |
Monoclonal | |
Protein A or G purified | |
Cancer, GPCR, Signal Transduction | |
Bovine,Human,Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title