Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTGES2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598099
Description
PTGES2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The PTGES2 gene encodes for the enzyme prostaglandin E synthase 2 (PTGES2), which is crucial for the biosynthesis of prostaglandin E2 (PGE2) from arachidonic acid. This enzyme operates downstream of the cyclooxygenase (COX) pathway, particularly interacting with COX-1 and COX-2 enzymes. PTGES2 is involved in various physiological and pathological processes by generating PGE2, which functions in inflammation, fever generation, and pain. Notably, PTGES2 is a membrane-associated protein and is distinct in its role compared to other prostaglandin synthases due to its specific functional and regulatory properties. Its activity is essential in fine-tuning the levels of PGE2 in tissues, impacting numerous signaling cascades, including those involved in immune responses and vascular functions.
Specifications
PTGES2 | |
Polyclonal | |
Unconjugated | |
Ptges2 | |
0610038H10Rik; C79137; C9orf15; FLJ14038; gamma-interferon-activated transcriptional element-binding factor 1; GATE-binding factor 1; Gbf1; GBF-1; membrane-associated prostaglandin E synthase 2; membrane-associated prostaglandin E synthase-2; MGC11289; Microsomal prostaglandin E synthase 2; microsomal prostaglandin E synthase-2; mPGE synthase-2; Mpges2; mPGES-2; PGES2; Prostaglandin E synthase 2; Prostaglandin E synthase 2 truncated form; Prostaglandin-H(2) E-isomerase; PTGES2; RP11-395P17.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80142 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H7Z7 | |
Ptges2 | |
Recombinant Human Prostaglandin E synthase 2 protein (88-377AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction