Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ PTGS2 (COX-2) Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human PTGS2 (NP_000954.1) (Met 1-Leu 604) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Sino Biological™ 12036H08B25

 View more versions of this product

Catalog No. 12-036-H08B25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human PTGS2 (NP_000954.1) (Met 1-Leu 604) fused to a polyhistidine tag at the C-terminus

  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 10% glycerol, pH 8.0
Description & Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
5743
≥90% by SDS-PAGE
Human
<1 EU/μg
PTGS2
Other proteins
PTGS2 (COX-2) Recombinant Human Protein
Lyophilized
68.5kd
5 x 5μg
Full Length, Recombinant
Prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase)
Assay Standard
Insect
His Tag
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit