Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGS2 Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB30313

Catalog No. 89-032-657


Add to cart

Description

Description

Rabbit polyclonal antibody raised against partial recombinant human PTGS2.

  • High specificity
  • High reactivity
Specifications

Specifications

PTGS2
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) The optimal working dilution should be determined by the end user.
P35354
COX-2|COX2|GRIPGHS|PGG/HS|PGHS-2|PHS-2|hCox-2
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against partial recombinant human PTGS2.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Liquid
PTGS2
Recombinant protein corresponding to human PTGS2.
Antigen affinity purification
RUO
Primary
5743
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)