Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PTH Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794115PLATES

 View more versions of this product

Catalog No. NB009862


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human PTH.
Specifications

Specifications

HRP
Sandwich ELISA
312.5 pg/mL
5741
RUO
PTH
Apoptosis, Biologically Active Proteins, Cancer
Human
PTH
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20000 pg/mL is recommended.
This solid phase sandwich ELISA utilizes an antibody specific for Human PTH.
Antibody Pairs
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human PTH polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.2 mg/mL of mouse anti-Human PTH monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 380 ng of recombinant Human PTH.
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    312.5-20000 pg/mL
    Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit