Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTH Mouse anti-Human, Clone: PTH/911, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14404
Description
Mouse monoclonal antibody raised against synthetic peptide of human PTH.
- High specificity
- High reactivity
Specifications
PTH | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
P01270 | |
PTH | |
A synthetic peptide corresponding to N-terminus of human PTH. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
PTH/911 | |
Mouse monoclonal antibody raised against synthetic peptide of human PTH. | |
In 10 mM PBS. | |
PTH1 | |
Mouse | |
Protein A/G purification | |
RUO | |
5741 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction