Learn More
PTH Mouse anti-Human, Clone: SPM604, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human PTH.
Manufacturer:  Abnova Corporation MAB13147
Catalog No. 89-934-807
Add to cart
Description
Description
The protein encoded by this gene is a hormone secreted by parathyroid cells. This hormone elevates blood Ca2+ level by dissolving the salts in bone and preventing their renal excretion. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH). [provided by RefSeq]
Specifications
Specifications
PTH
SPM604
Mouse monoclonal antibody raised against synthetic peptide of human PTH.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
PTH1
Mouse
IgG2b, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
PTH
A synthetic peptide corresponding to N-terminus region of human PTH.
Protein A/G purification
RUO
Primary
5741