Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTH Mouse anti-Human, Janelia Fluor 646, Clone: BGN/1F8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065676JF646
Description
PTH Monoclonal specifically detects PTH in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
PTH | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PTH | |
Synthetic peptide corresponding to amino acids 1-34 of mature PTH conjugated to a proprietary carrier molecule | |
0.1 mL | |
Primary | |
2 x 10^7with 1-34 PTH, 5 x 10^8 with 1-84 PTH | |
Store at 4°C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
BGN/1F8 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
Mouse | |
Protein A purified | |
Apoptosis, Biologically Active Proteins, Cancer | |
5741 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title