Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH Rabbit anti-Human, Allophycocyanin/Cy7, Clone: PTH/1717R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254474APCCY7

Catalog No. N254474APC7


Add to cart

Description

Description

PTH Monoclonal antibody specifically detects PTH in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

PTH
PTH/1717R
Flow Cytometry
Conjugated, Purified
PTH
Recombinant human PTH protein N-terminal fragment (exact sequence is proprietary)
Protein A or G purified
Apoptosis, Biologically Active Proteins, Cancer
Primary
5741.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit