Sino Biological™ PTH Recombinant Human Protein, N-GST Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human PTH (Ser 32-Phe 65) fused to GST tag at N-terminus

Manufacturer: Sino Biological™ 13192H09E50

 View more versions of this product

Catalog No. 13-192-H09E50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 31kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Human
    Structural Form Truncated, Recombinant
    Common Name Parathyroid Hormone
    Protein Tag GST Tag
    Validated Application Assay Standard
    Protein Family Hormones and Receptors
    Product Type PTH Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Gene ID (Entrez) 5741
    Endotoxin Concentration <1 EU/μg
    Gene Symbol PTH
    Expression System E.coli
    Format Lyophilized

    Expressed from DNA sequence encoding human PTH (Ser 32-Phe 65) fused to GST tag at N-terminus

    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit