Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH1R/PTHR1 Mouse anti-Human, Clone: 542407, R&D Systems™

Mouse Monoclonal Antibody

$116.82 - $341.00

Specifications

Antigen PTH1R/PTHR1
Clone 542407
Dilution Immunohistochemistry 8-25 ug/mL
Applications Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB57091100 View Documents R&D Systems
MAB57091100
100 ug Each for $341.00
Add to cart
 
MAB57091SP View Documents R&D Systems
MAB57091SP
25 ug Each for $116.82
Add to cart
 
Description

Description

PTH1R/PTHR1 Monoclonal antibody specifically detects PTH1R/PTHR1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

PTH1R/PTHR1
Immunohistochemistry 8-25 ug/mL
Unconjugated
Mouse
Human
Q03431
5745
IgG2a
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human PTHR1 in direct ELISAs.
542407
Immunohistochemistry
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHR, PTHR1, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor
Chinese hamster ovary cell line CHO-derived recombinant human PTHR1 Tyr23-Met189 Accession # Q03431
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit