Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCHD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548733

Catalog No. PA548733

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in neuronal proliferation and differentiation (PubMed:25281927). Plays a role in the accumulation of cellular cholesterol (). Involved in intracellular lipid droplet formation (PubMed:25281927). May contribute to cholesterol homeostasis in neuronal cells ().
Specifications

Specifications

PTCHD2
Polyclonal
PBS with 0.09% sodium azide
Q9P2K9
Patched domain-containing protein 2, Protein dispatched homolog 3, PTCHD2, DISP3, KIAA1337
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57540
Western Blot
Unconjugated
DISP3
Liquid
DISP3
KLH conjugated synthetic peptide between 179-207 amino acids from the N-terminal region of human PTHD2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit