Learn More
MBL International Corporation PaRathyroid Hormone Receptor 1 (PTHR1) Polyclonal Antibody (US Only)
Supplier:  MBL International Corporation MC761
Encompass_Preferred
Clonality:Polyclonal; Application:IHC; Conjugate:Unlabeled; Species Reactivity:Human; Size:50 µl; Target:PTHR1; PTHR1 Primary Antibody of 50 µl. For sale In the US only.PTHR1 Primary Antibody.
Catalog No. 50-492-30