Learn More
Tocris Bioscience™ PTIQ
Potent MMP-3 expression inhibitor; neuroprotective
Manufacturer:  Tocris Bioscience™ 4604/50
 View more versions of this product
Catalog No. 46-045-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent inhibitor of MMP-3 expression (IC50 = 60 nM); down-regulates induction of MMP-3 in both stressed dopaminergic cells and activated microglia. Also suppresses proinflammatory responses and inhibits NO production in activated microglia. Attenuates motor deficits, prevents neurodegeneration and suppresses microglial activation in a Parkinson's disease mouse model. Brain penetrant.
Specifications
Specifications
PTIQ
>98
50mg
PTIQ
235.28