Tocris™ PTIQ

Potent MMP-3 expression inhibitor; neuroprotective

Manufacturer: Tocris™ 4604/10

 View more versions of this product

Catalog No. 46-041-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors PTIQ
    Purity >98
    Molecular Weight 235.28
    Quantity 10mg
    Product Type PTIQ

    Potent inhibitor of MMP-3 expression (IC50 = 60nM); down-regulates induction of MMP-3 in both stressed dopaminergic cells and activated microglia. Also suppresses proinflammatory responses and inhibits NO production in activated microglia. Attenuates motor deficits, prevents neurodegeneration and suppresses microglial activation in a Parkinson's disease mouse model. Brain penetrant.

    • Chemical Name: 1-(3,4-Dihydro-7-hydroxy-6-methoxy-2(1H)-isoquinolinyl)-1-propanone
    • Formula: C13H17NO3
    • CAS Number: 1032822-42-6
    • Solubility: Soluble to 100mM in DMSO and to 20mM in 1eq. NaOH
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit