Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK9 Mouse anti-Human, Clone: OTI5C12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527077

Catalog No. PIMA527077


Add to cart

Description

Description

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
Specifications

Specifications

PTK9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q12792
A6, PTK9, A6 protein tyrosine kinase, PTK9 protein tyrosine kinase 9, protein A6, protein tyrosine kinase 9, twinfilin-1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5756
Immunohistochemistry (Paraffin), Western Blot
OTI5C12
Unconjugated
TWF1
Liquid
TWF1
Human recombit protein fragment corresponding to amino acids 140-384 of TWF1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit