Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human PTN Full-length ORF (NP_002816, 1 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00005764P01
Description
Sequence: MQAQQYQQQRRKFAAAFLAFIFILAAVDTAEAGKKEKPEKKVKKSDCGEWQWSVCVPTSGDCGLGTREGTRTGAECKQTMKTQRCKIPCNWKKQFGAECKYQFQAWGECDLNTALKTRTGSLKRALHNAECQKTVTISKPCGKLTKPKPQAESKKKKKEGKKQEKMLDSpecifications
NP_002816 | |
wheat germ expression system | |
Liquid | |
HARP|HBGF8|HBNF|NEGF1 | |
PTN | |
MQAQQYQQQRRKFAAAFLAFIFILAAVDTAEAGKKEKPEKKVKKSDCGEWQWSVCVPTSGDCGLGTREGTRTGAECKQTMKTQRCKIPCNWKKQFGAECKYQFQAWGECDLNTALKTRTGSLKRALHNAECQKTVTISKPCGKLTKPKPQAESKKKKKEGKKQEKMLD | |
PTN (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
RUO |
PTN | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
5764 | |
Wheat Germ (in vitro) | |
44.11kDa | |
GST | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title