Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTOV1 Polyclonal Antibody
PTOV1 Polyclonal Antibody
Supplier: Thermo Scientific PA567569
Description
PTOV1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
PTOV1 | |
Polyclonal | |
Unconjugated | |
PTOV1 | |
1110030G05Rik; ACID2; Activator interaction domain-containing protein 2; AU041779; Gcap3; granule cell antiserum positive 3; PP642; prostate tumor over expressed gene 1; prostate tumor overexpressed 1; prostate tumor-overexpressed gene 1 protein; Prostate tumor-overexpressed gene 1 protein homolog; Ptov1; PTOV-1; UNQ6127/PRO20092 | |
Rabbit | |
affinity chromatography | |
RUO | |
292888, 53635, 84113 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q5U2W6, Q86YD1, Q91VU8 | |
PTOV1 | |
Synthesized peptide derived from human PTOV1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction