Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP alpha/PTPRA Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300087IR

Catalog No. NB064356


Add to cart

Description

Description

PTP alpha/PTPRA Polyclonal antibody specifically detects PTP alpha/PTPRA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PTP alpha/PTPRA
DyLight 755
50mM Sodium Borate
EC 3.1.3.48, HEPTP, LRPHPTPalpha, protein tyrosine phosphatase, receptor type, A, Protein-tyrosine phosphatase alpha, PTPAreceptor type, alpha polypeptide, PTPLCA-related phosphatase, receptor-type tyrosine-protein phosphatase alpha, RPTPA, R-PTP-alpha, tyrosine phosphatase alpha
Produced in rabbits immunized with purified, recombinant PTP alpha/PTPRA (P18433-2; Ala174-Lys793).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
GPCR, Phospho Specific, Protein Phosphatase, Signal Transduction
Primary
5786
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit