Learn More
PTPD1 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS19587R
Catalog No. 50-198-4346
Add to cart
Description
Description
The protein tyrosine phosphatase (PTP) family of proteins are signaling molecules that regulate processes such as cell growth, cell differentiation, oncogenic transformation and the mitotic cycle. PTPN21 (protein tyrosine phosphatase, non-receptor type 21), also known as PTPD1 or PTPRL10, is a 1,174 amino acid member of the PTP family and localizes to both the cytoplasm and the cytoskeleton. Containing one FERM domain and one tyrosine-protein phosphatase domain, PTPN21 functions to catalyze the water-dependent conversion of a protein tyrosine phosphate into a protein tyrosine and a free phosphate and, via this catalytic activity, may be involved in liver regeneration and spermatogenesis. The gene encoding PTPN21 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome.
Specifications
Specifications
PTPD1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTPN21
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
11099
PTPN21
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q16825
dJ1175B15.2 (protein tyrosine phosphatase, non-receptor type 21); protein tyrosine phosphatase 2E; protein tyrosine phosphatase, non-receptor type 21; Protein-tyrosine phosphatase 2E; Protein-tyrosine phosphatase D1; protein-tyrosine phosphatase PTP-RL10; Ptp2e; PTPD1; PTPN21; PTPRL10; Tyrosine-protein phosphatase non-receptor type 21
KLH conjugated synthetic peptide derived from human PTPD1.
Protein A
RUO
Antibody
Polyclonal
Human