Learn More
PTPIP51 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206411AP
Catalog No. 50-173-3009
Add to cart
Description
Description
RMDN3 is a protein coding gene involved in cellular calcium homeostasis regulation. May participate in differentiation and apoptosis of keratinocytes. Overexpression induces apoptosis.
Specifications
Specifications
PTPIP51
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96TC7
FAM82A2, FAM82C, hRMD 3, Protein FAM82A2, Protein FAM82C, PTPIP51, RMD 3, RMD3
Rabbit
IgG
150 μL
-20°C
Primary
55177
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RMDN3
Liquid
RMDN3
PTPIP51 Fusion Protein Ag14708
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human