Learn More
Aviva Systems Biology Corp HACD3 Antibody - N-terminal region (ARP47251_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTPLAD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PTPLAD1. Alternate names: B-IND1, FLJ90376, HSPC121, HACD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59498