Learn More
Invitrogen™ PTPLAD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113839
Description
Immunogen sequence: WLDESDAEME LRAKEEERLN KLRLESEGSP ETLTNLRKGY LFMYNLVQFL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Catalyzes the third of the four reactions of the long-chain fatty acids elongation cycle. This endoplasmic reticulum-bound enzymatic process, allows the addition of two carbons to the chain of long- and very long-chain fatty acids/VLCFAs per cycle. This enzyme catalyzes the dehydration of the 3-hydroxyacyl-CoA intermediate into trans-2,3-enoyl-CoA, within each cycle of fatty acid elongation. Thereby, it participates in the production of VLCFAs of different chain lengths that are involved in multiple biological processes as precursors of membrane lipids and lipid mediators. May be involved in Rac1-signaling pathways leading to the modulation of gene expression. Promotes insulin receptor/INSR autophosphorylation and is involved in INSR internalization (PubMed:25687571).
Specifications
PTPLAD1 | |
Polyclonal | |
Unconjugated | |
HACD3 | |
3-hydroxyacyl-CoA dehydratase 3; BIND1; B-ind1; Butyrate-induced protein 1; butyrate-induced transcript 1; HACD3; HSPC121; protein tyrosine phosphatase-like A domain containing 1; protein tyrosine phosphatase-like protein PTPLAD1; protein-tyrosine phosphatase-like A domain-containing protein 1; PTPLAD1; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 3; very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51495 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9P035 | |
HACD3 | |
Synthetic peptide directed towards the N terminal region of human PTPLAD1 (aa 110-159). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.