Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPLAD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553239
Description
Immunogen sequence: MENQVLTPHV YWAQRHRELY LRVELSDVQN PAISITENVL HFKAQGHGAK GDNVYEFHLE FLDLVKPEPV YKLTQRQVNI TVQKKVSQWW ERLTKQEKRP LFLAPDFDRW LDESDAEMEL RAKEEERLNK LRLE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Catalyzes the third of the four reactions of the long-chain fatty acids elongation cycle. This endoplasmic reticulum-bound enzymatic process, allows the addition of two carbons to the chain of long- and very long-chain fatty acids/VLCFAs per cycle. This enzyme catalyzes the dehydration of the 3-hydroxyacyl-CoA intermediate into trans-2,3-enoyl-CoA, within each cycle of fatty acid elongation. Thereby, it participates in the production of VLCFAs of different chain lengths that are involved in multiple biological processes as precursors of membrane lipids and lipid mediators. May be involved in Rac1-signaling pathways leading to the modulation of gene expression. Promotes insulin receptor/INSR autophosphorylation and is involved in INSR internalization (PubMed:25687571).
Specifications
PTPLAD1 | |
Polyclonal | |
Unconjugated | |
HACD3 | |
3-hydroxyacyl-CoA dehydratase 3; BIND1; B-ind1; Butyrate-induced protein 1; butyrate-induced transcript 1; HACD3; HSPC121; protein tyrosine phosphatase-like A domain containing 1; protein tyrosine phosphatase-like protein PTPLAD1; protein-tyrosine phosphatase-like A domain-containing protein 1; PTPLAD1; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 3; very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51495 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P035 | |
HACD3 | |
Recombinant protein corresponding to Human PTPLAD1. Recombinant protein control fragment (Product #RP-91208). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction