Sino Biological™ PTPMT1 Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human PTPMT1 isoform 1 (Lys 28-Thr 201) fused to polyhistidine tag at N-terminus

Manufacturer: Sino Biological™ 12902H07E50

 View more versions of this product

Catalog No. 12-902-H07E50

  N/A
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 21.7kd
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Source Human
    Common Name Protein Tyrosine Phosphatase, Mitochondrial 1
    Gene Symbol PTPMT1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System E.coli
    Product Type PTPMT1 Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 114971

    Expressed from DNA sequence encoding human PTPMT1 isoform 1 (Lys 28-Thr 201) fused to polyhistidine tag at N-terminus

    • Formulation: PBS, 10% Glycerol, 1mM DTT, pH 7.4

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit