Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPmu/PTPRM Mouse anti-Human, Clone: SBK10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266638
Description
PTPmu/PTPRM Monoclonal antibody specifically detects PTPmu/PTPRM in Human samples. It is validated for Western Blot.Specifications
PTPmu/PTPRM | |
SBK10 | |
Western Blot | |
Purified | |
PTPRM | |
Proprietary | |
Protein A or G purified | |
Protein Phosphatase | |
Primary | |
5797 |
Western Blot | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
EC 3.1.3.48, hR-PTPu, MGC166994, protein tyrosine phosphatase mu, protein tyrosine phosphatase, receptor type, M, protein tyrosine phosphatase, receptor type, mu polypeptide, Protein-tyrosine phosphatase mu, PTPRL1R-PTP-MU, receptor-type tyrosine-protein phosphatase mu, RPTPM, R-PTP-mu, RPTPU | |
Mouse | |
IgG1 | |
100μg | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title