Learn More
PTPN1 Mouse anti-Canine, Human, Clone: OTI1D10, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA503310
Catalog No. 50-167-4237
Add to cart
Description
Description
Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (~450 kDa) consisting of 24 subunits of two types, H (heavy) and L (light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.
Specifications
Specifications
PTPN1
Monoclonal
Unconjugated
PTPN1
Liquid
PTPN1
Full length human recombit protein of human PTPN1 produced in HEK293 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D10
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P18031
PTP1B
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
477263, 5770