Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN1 Mouse anti-Human, Clone: OTI1A2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503038

Catalog No. 50-167-4012


Add to cart

Description

Description

Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (~450 kDa) consisting of 24 subunits of two types, H (heavy) and L (light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.
Specifications

Specifications

PTPN1
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P18031
PTP1B
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5770
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1A2
Unconjugated
PTPN1
Liquid
PTPN1
Full length human recombit protein of human PTPN1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit