Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PTPN11, Rabbit anti-Human, Clone: 4A2, Abnova™

Rabbit Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB19077

Catalog No. 89-335-911



Specifications

Specifications

PTPN11
4A2
Flow Cytometry (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user.
protein tyrosine phosphatase, non-receptor type 11
BPTP3|CFC|MGC14433|NS1|PTP-1D|PTP2C|SH-PTP2|SH-PTP3|SHP2
Rabbit
IgG1k
RUO
Antibody
Monoclonal
Human
Flow Cytometry
Unconjugated
In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA).
Liquid
PTPN11
A synthetic phosphopeptide corresponding to residues surrounding Y580 of human PTPN11.
200 μL
Store at -20°C.
Primary
5781
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit