Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN12 Mouse anti-Human, Clone: AG25, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266639
Description
PTPN12 Monoclonal antibody specifically detects PTPN12 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PTPN12 | |
AG25 | |
Western Blot, Immunoprecipitation 1:10 - 1:500 | |
PBS with 0.08% Sodium Azide | |
5782 | |
Mouse | |
IgG1 | |
Primary | |
100μg | |
Human,Mouse |
Immunoprecipitation,Western Blot | |
Unconjugated | |
Purified | |
protein tyrosine phosphatase, non-receptor type 12, Protein-tyrosine phosphatase G1, PTPG1tyrosine-protein phosphatase non-receptor type 12, PTP-PESTEC 3.1.3.48 | |
PTPN12 | |
baculovirus expressed, human full length PTP-PEST | |
Monoclonal | |
Protein A or G purified | |
Store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title