Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPN13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139860
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: STPVHLTNEM KNYMKKSSYM QDSAIDSSSK DHHWSRGTLR HISENSFGPS GGLREGSLSS QDSRTESASL SQSQVNGFFA SHLGDQTWQE SQHGSPSPSV ISKATE.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported.
Specifications
PTPN13 | |
Polyclonal | |
Unconjugated | |
PTPN13 | |
AI324989; DKFZp686J1497; FAP-1; Fas-associated protein-tyrosine phosphatase 1; hPTP1E; hPTPE1; PNP1; protein Tyr phosphatase; protein tyrosine phosphatase DPZPTP; Protein tyrosine phosphatase PTP-BL; protein tyrosine phosphatase, non-receptor type 13; protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associated phosphatase); protein-tyrosine phosphatase 1E; Protein-tyrosine phosphatase PTPL1; protein-tyrosine phosphatase RIP; ptp; Ptp14; PTP1E; PTP36; PTP-BAS; PTP-BL; PTP-E1; PTPL1; PTPLE; PTPN13; Ptpri; RIP; tyrosine phosphatase; tyrosine-protein phosphatase non-receptor type 13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12923 | |
PTPN13 | |
Recombinant protein corresponding to Human PTPN13. Recombinant protein control fragment (Product #RP-108381). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction