Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN20A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548277

Catalog No. PA548277

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Tyrosine-protein phosphatase targeted to sites of actin polymerization in response of varied extracellular stimuli. Has tyrosine phosphatase activity towards various tyrosyl phosphorylated substrates.
Specifications

Specifications

PTPN20A
Polyclonal
PBS with 0.09% sodium azide
Q4JDL3
Tyrosine-protein phosphatase non-receptor type 20, hPTPN20, PTPN20A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26095
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTPN20
Liquid
PTPN20
KLH conjugated synthetic peptide between 178-207 amino acids from the Central region of human PTPN20A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit