Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPN23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576478
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Plays a role in sorting of endocytic ubiquitinated cargos into multivesicular bodies (MVBs) via its interaction with the ESCRT-I complex (endosomal sorting complex required for transport I), and possibly also other ESCRT complexes. May act as a negative regulator of Ras-mediated mitogenic activity. Plays a role in ciliogenesis.
Specifications
PTPN23 | |
Polyclonal | |
Unconjugated | |
PTPN23 | |
AI462446; HDPTP; HD-PTP; His domain-containing protein tyrosine phosphatase; Kiaa1471; Protein tyrosine phosphatase TD14; protein tyrosine phosphatase, non-receptor type 23; PTPN23; PTP-TD14; Tyrosine-protein phosphatase non-receptor type 23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
104831, 117552, 25930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O88902, Q6PB44, Q9H3S7 | |
PTPN23 | |
Recombinant fusion protein of human PTPN23. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction