Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ PTPN4 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$344.00 - $976.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris buffer (pH 8.0) containing 10% glycerol |
Gene ID (Entrez) | 5775 |
Molecular Weight | TMW: 32kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2520101
|
Novus Biologicals™
NBP2520110.1MG |
0.1mg |
Each for $344.00
|
|
NBP2520105
|
Novus Biologicals™
NBP2520110.5MG |
0.5mg |
Each for $976.00
|
|
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 655-913 of Human PTPN4 The Recombinant Human PTPN4 Protein is derived from E. coli. The Recombinant Human PTPN4 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
20mM Tris buffer (pH 8.0) containing 10% glycerol | |
TMW: 32kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
PTPN4 | |
Yes |
SDS-Page | |
5775 | |
>85%, by SDS-PAGE | |
EC 3.1.3.48, MEG, megakaryocyte phosphatase, megakaryocyte protein-tyrosine phosphatase, protein tyrosine phosphatase MEG1, protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte), Protein-tyrosine phosphatase MEG1, PTPase-MEG1, PTPMEG, PTPMEG1, tyrosine-protein phosphatase non-receptor type 4 | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title