Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN7 Rabbit anti-Human, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP304491100UL
Description
PTPN7 Polyclonal antibody specifically detects PTPN7 in Human samples. It is validated for Western BlotSpecifications
PTPN7 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
100 μL | |
Cancer | |
Primary | |
5778 |
Western Blot | |
PBS with 50% glycerol, pH7.3. | |
BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human PTPN7 (NP_542155.1). | |
Affinity purified | |
RUO | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title