Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30449120UL

 View more versions of this product

Catalog No. NB095966


Add to cart

Description

Description

PTPN7 Polyclonal antibody specifically detects PTPN7 in Human samples. It is validated for Western Blot
Specifications

Specifications

PTPN7
Unconjugated
Purified
Rabbit
IgG
20 μL
Cancer
Primary
5778
Western Blot
PBS with 50% glycerol, pH7.3.
BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human PTPN7 (NP_542155.1).
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit