Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen PTPN7
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095965
View Documents
Novus Biologicals
NBP304491100UL
100 μg Each for $320.71
Add to cart
 
NB095966
View Documents
Novus Biologicals
NBP30449120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

PTPN7 Polyclonal antibody specifically detects PTPN7 in Human samples. It is validated for Western Blot
Specifications

Specifications

PTPN7
Unconjugated
Rabbit
Cancer
PBS with 50% glycerol, pH7.3.
5778
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human
BPTP-4, FLJ18703, HEPTPnonreceptor-type, stress induced, LPTP, protein tyrosine phosphatase, non-receptor type 7, tyrosine-protein phosphatase non-receptor type 7
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human PTPN7 (NP_542155.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit