Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN7 Mouse anti-Human, Clone: OTI3G2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525701

Catalog No. PIMA525701


Add to cart

Description

Description

The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This gene is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The noncatalytic N-terminus of this PTP can interact with MAP kinases and suppress the MAP kinase activities. This PTP was shown to be involved in the regulation of T cell antigen receptor signaling, which was thought to function through dephosphorylating the molecules related to MAP kinase pathway. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

PTPN7
Monoclonal
1.02 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P35236
Tyrosine-protein phosphatase non-receptor type 7; Protein-tyrosine phosphatase LC-PTP; Hematopoietic protein-tyrosine phosphatase; LPTP; HEPTP; PTPNI; BPTP-4; LC-PTP
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5778
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3G2
Unconjugated
PTPN7
Liquid
PTPN7
Full length human recombit protein of PTPN7 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit