Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRC Mouse anti-Human, Clone: HI30 (CF-Blue), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13941

Catalog No. 89-034-006


Add to cart

Description

Description

Mouse monoclonal antibody raised against human PTPRC.

  • High specificity
  • High reactivity
Specifications

Specifications

PTPRC
HI30
Mouse monoclonal antibody raised against human PTPRC.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
PTPRC
Human PBMCs and tonsil cells.
Protein A/G purification
RUO
Primary
5788
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Flow Cytometry (5 uL/106 cells) Functional Study Immunocytochemistry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot The optimal w
P08575
B220|CD45|CD45R|GP180|LCA|LY5|T200
Mouse
IgG1
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)