Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRC Mouse anti-Human, Clone: PTPRC/1461, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14697

Catalog No. 89-034-780


Add to cart

Description

Description

Mouse monoclonal antibody raised against human PTPRC.

  • High specificity
  • High reactivity
Specifications

Specifications

PTPRC
PTPRC/1461
Mouse monoclonal antibody raised against human PTPRC.
In 10 mM PBS (0.05% BSA and 0.05% azide).
B220|CD45|CD45R|GP180|LCA|LY5|T200
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
PTPRC
Human B-lymphocytes.
Protein A/G purification
RUO
Primary
5788
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)