Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRC, Mouse anti-Human, FITC, Clone: F10-89-4, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16148

Catalog No. 89-998-922


Add to cart

Description

Description

Specifications

Specifications

PTPRC
F10-89-4
Flow Cytometry (1:1) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
protein tyrosine phosphatase, receptor type, C
B220/CD45/CD45R/GP180/LCA/LY5/T200
Mouse
IgG2a
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin)
FITC
In PBS, pH 7.4 (1% BSA, 0.09% sodium azide).
Liquid
PTPRC
Human T lymphocytes.
50 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
5788
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit