Learn More
PTPRC Rat anti-Human, Clone: YTH24.5, Abnova™
Rat monoclonal antibody raised against human PTPRC.
Manufacturer:  Abnova Corporation MAB12737
Catalog No. 89-934-826
Add to cart
Description
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus belongs to receptor type PTP. This gene is specifically expressed in hematopoietic cells. This PTP has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. It functions through either direct interaction with components of the antigen receptor complexes, or by activating various Src family kinases required for the antigen receptor signaling. This PTP also suppresses JAK kinases, and thus functions as a regulator of cytokine receptor signaling. Four alternatively spliced transcripts variants of this gene, which encode distinct isoforms, have been reported. [provided by RefSeq]
Specifications
Specifications
PTPRC
YTH24.5
Rat monoclonal antibody raised against human PTPRC.
In PBS (0.09% sodium azide).
B220/CD45/CD45R/GP180/LCA/LY5/T200
Rat
IgG2b
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (1:10-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
PTPRC
Human T lymphocytes
Protein A purification
RUO
Primary
5788