Learn More
PTPRCAP / CD45AP Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 197151AP
Catalog No. 50-173-2505
Add to cart
Description
Description
MSK-1 is a mitogen and stress activated protein kinase-1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK-1 can be activated by ERK1/2 and SAPK2 /p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG-14 in response to mitogen and stress. Similar to RSK, MSK-1 contains two kinase domains (N-term and a C-term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK-1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C-terminal kinase domain of MSK-1 which is essential for the catalytic activity of the N-terminal kinase domain.
Specifications
Specifications
PTPRCAP / CD45AP
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14761
CD45 AP, CD45 associated protein, LPAP, PTPRC associated protein, PTPRCAP
Rabbit
IgG
150 μL
-20°C
Primary
5790
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTPRCAP
Liquid
PTPRCAP
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human