Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPRCAP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548784

Catalog No. PA548784

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene was identified as a transmembrane phosphoprotein specifically associated with tyrosine phosphatase PTPRC/CD45, a key regulator of T- and B-lymphocyte activation. The interaction with PTPRC may be required for the stable expression of this protein. [provided by RefSeq, Jul 2008]
Specifications

Specifications

PTPRCAP
Polyclonal
PBS with 0.09% sodium azide
Q14761
Protein tyrosine phosphatase receptor type C-associated protein, PTPRC-associated protein, CD45-associated protein, CD45-AP, Lymphocyte phosphatase-associated phosphoprotein, PTPRCAP, LPAP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5790
Western Blot
Unconjugated
PTPRCAP
Liquid
PTPRCAP
KLH conjugated synthetic peptide between 90-118 amino acids from the Central region of human PTPRCAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit