Learn More
PTPRD Rat anti-Mouse, Unconjugated, Clone: F34a6, MilliporeSigma™
Rat Monoclonal Antibody
Manufacturer:  MilliporeSigma MABS218925UG
 View more versions of this product
Catalog No. MABS21892MI
Add to cart
Description
Description
PTPRD Monoclonal antibody specifically detects PTPRD in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
PTPRD
F34a6
NP_035341.2
Receptor-type tyrosine-protein phosphatase delta;EC:3.1.3.48;Protein-tyrosine phosphatase delta;R-PTP-delta
Rat
IgG2bκ
Protein G Purified
Signaling
Primary
Clone F34a6 is a rat monoclonal antibody that specifically detects different isoforms of murine Receptor-type tyrosine-protein phosphatase delta (PTPRD), but does not react with LAR. It targets an epitope within the ectodomain region.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Purified
Ptprd
GST-tagged recombinant fragments corresponding to Fibronectin type III repeats 2,3 and 8 of murine Receptor-type tyrosine-protein phosphatase delta (PTPRD).
∼80, 120, 130 kDa observed, 214.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μg
Stable for 1 year at 2 to 8°C from date of receipt.
Monoclonal
Mouse