Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPRQ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562764
Description
Immunogen sequence: AVITGPTCYL IDVKSVDNDE FNISFIKSNE ENKTIEIKDL EIFTRYSVVI TAFTGNISAA YVEGKSSAEM IVTTLESAPK DPPNNMTFQK IPDEVTK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
Protein Tyrosine Phosphatase RQ.
Specifications
PTPRQ | |
Polyclonal | |
Unconjugated | |
PTPRQ | |
DFNB84; DFNB84A; phosphatidylinositol phosphatase PTPRQ; phosphotidylinositol phosphatase PTPRQ; protein tyrosine phosphatase, receptor type Q; protein tyrosine phosphatase, receptor type, Q; protein-tyrosine phosphatase, receptor-type, expressed by glomerular mesangial cells; PTPGMC1; PTPRQ; R-PTP-Q | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
374462 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UMZ3 | |
PTPRQ | |
Recombinant protein corresponding to Human PTPRQ. Recombinant protein control fragment (Product #RP-101329). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction